2,3,4,5-Tetrafluorobenzoyl chloride ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,3,4,5-tetrafluorobenzoylchloride

T1855-25G T1855-5G
TCT1855-25GEA 45 USD
TCT1855-25G TCT1855-5G
2,3,4,5-Tetrafluorobenzoyl chloride ≥97.0% (by GC, titration analysis)
2,3,4,5-Tetrafluorobenzoyl chloride

CAS Number: 94695-48-4
MDL Number: MFCD00075164
Molecular Formula: C7HClF4O
Molecular Weight: 212.53
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 89
Specific Gravity (20/20): 1.61

Formula: C₇HClF₄O
MW: 212.53 g/mol
Density: 1.61 (20 °C)
Flash Pt: 89 °C
MDL Number: MFCD00075164
CAS Number: 94695-48-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9e9fe30f2303d487ba0d5d43b4284d03f97741a&vid=d9e9fe30f2303d487ba0d5d43b4284d03f97741a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR