Tetrahydro-4H-pyran-4-one ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Dihydro-2H-pyran-4(3H)-one, Tetrahydro-α-pyran-4-one, Tetrahydro-2H-pyran-4-one, Tetrahydro-γ-pyran-4-one

T1806-1G T1806-25G T1806-5G
TCT1806-1GEA 36 USD
TCT1806-1G TCT1806-25G TCT1806-5G
Tetrahydro-4H-pyran-4-one ≥95.0% (by GC)
Tetrahydro-4H-pyran-4-one

CAS Number: 29943-42-8
MDL Number: MFCD00006581
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 69
Flash Point (°C): 55
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: 4-Oxotetrahydropyran

Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 69 °C
Density: 1.09 (20 °C)
Flash Pt: 55 °C
MDL Number: MFCD00006581
CAS Number: 29943-42-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac48150a6f6cbe06105132766183bd5847b95248&vid=ac48150a6f6cbe06105132766183bd5847b95248&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR