4-(Trifluoromethyl)benzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T1795-1G T1795-5G
TCT1795-1GEA 28 USD
TCT1795-1G TCT1795-5G
4-(Trifluoromethyl)benzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
4-(Trifluoromethyl)benzenesulphonyl chloride

CAS Number: 2991-42-6
MDL Number: MFCD00042422
Molecular Formula: C7H4ClF3O2S
Molecular Weight: 244.61
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 58
Flash Point (°C): 110
Freezing point (°C): 26


Other Product Names / Synonyms: alpha,alpha,alpha-Trifluorotoluene-4-sulfonyl Chloride

Formula: C₇H₄ClF₃O₂S
Boiling Pt: 58 °C
Flash Pt: 110 °C
MDL Number: MFCD00042422
CAS Number: 2991-42-6
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b8b41fe5e3a2cc9ad0e794c7573401fe2019e65&vid=2b8b41fe5e3a2cc9ad0e794c7573401fe2019e65&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR