α-Bromo-4-(trifluoromethoxy)toluene ≥97.0% (by GC)

Supplier: TCI America

T1781-1G T1781-5G
TCT1781-1GEA 35.62 USD
TCT1781-1G TCT1781-5G
α-Bromo-4-(trifluoromethoxy)toluene ≥97.0% (by GC)
α-Bromo-4-(trifluoromethoxy)toluene

CAS Number: 50824-05-0
MDL Number: MFCD00061238
Molecular Formula: C8H6BrF3O
Molecular Weight: 255.03
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 79
Flash Point (°C): 94


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha-Bromo-4-(trifluoromethoxy)toluene

Formula: C₈H₆BrF₃O
MW: 255.03 g/mol
Boiling Pt: 79 °C
Flash Pt: 94 °C
MDL Number: MFCD00061238
CAS Number: 50824-05-0
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c8eb94d770cc7c7be6a28450e08498515ecc7b8&vid=0c8eb94d770cc7c7be6a28450e08498515ecc7b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR