Allyl trifluoroacetate ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Allyl Trifluoroacetate, ['Trifluoroacetic acid Allyl Ester'], Trifluorethansäure(2-propenyl)ester, Trifluoroacetic allyl ester

T1765-25G T1765-5G
TCT1765-25GEA 195 USD
TCT1765-25G TCT1765-5G
Allyl trifluoroacetate ≥95.0% (by GC)
Allyl trifluoroacetate

CAS Number: 383-67-5
MDL Number: MFCD00013567
Molecular Formula: C5H5F3O2
Molecular Weight: 154.09
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 66
Flash Point (°C): -1
Specific Gravity (20/20): 1.18


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Trifluoroacetic Acid Allyl Ester

Formula: C₅H₅F₃O₂
MW: 154.09 g/mol
Boiling Pt: 66 °C
Density: 1.18 (20 °C)
Flash Pt: –1 °C
MDL Number: MFCD00013567
CAS Number: 383-67-5
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ae77c55516e78af1ff892beba37e3a603ff9b72&vid=0ae77c55516e78af1ff892beba37e3a603ff9b72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR