2,4,6-Trimethylbenzyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 1585-16-6
MDL Number: MFCD00000901
Molecular Formula: C10H13Cl
Molecular Weight: 168.66
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 119
Melting point (°C): 37
Flash Point (°C): 106
Storage Temperature: 0-10°C
Other Product Names / Synonyms: alpha2-Chloroisodurene, 2-Chloromethyl-1,3,5-trimethylbenzene
Formula:
C₁₀H₁₃Cl Boiling Pt: 119 °C Melting Pt: 37 °C Flash Pt: 106 °C |
MDL Number:
MFCD00000901 CAS Number: 1585-16-6 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...