(S)-(-)-Tetrahydro-2-furoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

T1741-1G T1741-25G T1741-5G
TCT1741-1GEA 17 USD
TCT1741-1G TCT1741-25G TCT1741-5G
(S)-(-)-Tetrahydro-2-furoic acid ≥97.0% (by GC, titration analysis)
(S)-(-)-Tetrahydro-2-furoic acid

CAS Number: 87392-07-2
MDL Number: MFCD00272639
Molecular Formula: C5H8O3
Molecular Weight: 116.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 251
Flash Point (°C): 139
Specific Gravity (20/20): 1.21
Specific rotation [a]20/D: -19 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-2-Tetrahydrofuroic Acid

Formula: C₅H₈O₃
Boiling Pt: 251 °C
Density: 1.21 (20 °C)
Flash Pt: 139 °C
MDL Number: MFCD00272639
CAS Number: 87392-07-2
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3eea230ddbe1b563ac429e115cafaac86db6b4b3&vid=3eea230ddbe1b563ac429e115cafaac86db6b4b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR