(R)-(+)-Tetrahydro-2-furoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T1740-25G T1740-5G
TCT1740-25GEA 380 USD
TCT1740-25G TCT1740-5G
(R)-(+)-Tetrahydro-2-furoic acid ≥98.0% (by GC, titration analysis)
(R)-(+)-Tetrahydro-2-furoic acid

CAS Number: 87392-05-0
MDL Number: MFCD00211271
Molecular Formula: C5H8O3
Molecular Weight: 116.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 129
Flash Point (°C): 139
Specific Gravity (20/20): 1.22
Specific rotation [a]20/D: 19 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(+)-2-Tetrahydrofuroic Acid

Formula: C₅H₈O₃
Boiling Pt: 129 °C
Density: 1.22 (20 °C)
Flash Pt: 139 °C
MDL Number: MFCD00211271
CAS Number: 87392-05-0
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb4195957a0989f45b4de936180b48a29089dbc2&vid=fb4195957a0989f45b4de936180b48a29089dbc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR