Triethylvinylsilane ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Vinyltriethylsilane

T1703-25ML T1703-5ML
TCT1703-25MLEA 259 USD
TCT1703-25ML TCT1703-5ML
Triethylvinylsilane ≥95.0% (by GC)
Triethylvinylsilane

CAS Number: 1112-54-5
MDL Number: MFCD00041345
Molecular Formula: C8H18Si
Molecular Weight: 142.32
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 147
Flash Point (°C): 25
Specific Gravity (20/20): 0.77


Product Categories: Synthetic Organic Chemistry, Vinylation
Other Product Names / Synonyms: Vinyltriethylsilane

Formula: C₈H₁₈Si
MW: 142.32 g/mol
Boiling Pt: 147 °C
Density: 0.77 (20 °C)
Flash Pt: 25 °C
MDL Number: MFCD00041345
CAS Number: 1112-54-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=318c9bcce9fb64643c90c1965a7e4d8f4a3e1eb5&vid=318c9bcce9fb64643c90c1965a7e4d8f4a3e1eb5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR