Tetrabutylammonium dihydrogentrifluoride ≥90.0% (by titrimetric analysis)

Supplier: TCI America

T1635-25G T1635-5G
TCT1635-25GEA 184 USD
TCT1635-25G TCT1635-5G
Tetrabutylammonium dihydrogentrifluoride ≥90.0% (by titrimetric analysis)
Tetrabutylammonium dihydrogentrifluoride

CAS Number: 99337-56-1
MDL Number: MFCD00145365
Molecular Formula: C16H38F3N
Molecular Weight: 301.48
Purity/Analysis Method: >90.0% (T)
Form: Clear Liquid
Color: Colorless

Formula: C₁₆H₃₈F₃N
MDL Number: MFCD00145365
CAS Number: 99337-56-1
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85a6fcfe45cbb373d144cd2631cd72250cd875b4&vid=85a6fcfe45cbb373d144cd2631cd72250cd875b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR