2,4,6-Triisopropylbenzenesulfonyl hydrazide ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,4,6-Triisopropylbenzenesulphonohydrazide

T1591-25G T1591-5G
TCT1591-25GEA 496 USD
TCT1591-25G TCT1591-5G
2,4,6-Triisopropylbenzenesulfonyl hydrazide ≥95.0% (by titrimetric analysis)
2,4,6-Triisopropylbenzenesulfonyl hydrazide

CAS Number: 39085-59-1 MDL Number: MFCD00014750 Molecular Formula: C15H26N2O2S Molecular Weight: 298.45 Purity/Analysis Method: 95.0% (T) Form: Crystal Color: White Melting point (°C): 122 Storage Temperature: 0°C

Formula: C₁₅H₂₆N₂O₂S
MW: 298.45 g/mol
Melting Pt: 122 °C
MDL Number: MFCD00014750
CAS Number: 39085-59-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2aab752a3167a8858dd8967fcf0116886b483b0&vid=c2aab752a3167a8858dd8967fcf0116886b483b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR