Triisopropylsilyl trifluoromethanesulphonate ≥98.0%

Supplier: TCI America

Synonyms: Trifluoromethanesulphonic acid triisopropylsilyl ester, Tri(propan-2-yl)silyl trifluoromethanesulfonate

T1588-25G T1588-5G
TCT1588-25GEA 122.59 USD
TCT1588-25G TCT1588-5G
Triisopropylsilyl trifluoromethanesulphonate ≥98.0%
Triisopropylsilyl trifluoromethanesulphonate

CAS Number: 80522-42-5
MDL Number: MFCD00009913
Molecular Formula: C10H21F3O3SSi
Molecular Weight: 306.42
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 87
Flash Point (°C): 100
Specific Gravity (20/20): 1.14


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Protection & Derivatization Reagents (for Synthesis), Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Triisopropylsilyl Triflate, Trifluoromethanesulfonic Acid Triisopropylsilyl Ester

Formula: C₁₀H₂₁F₃O₃SSi
MW: 306.42 g/mol
Boiling Pt: 120
Density: 1.14 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00009913
CAS Number: 80522-42-5
UN: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9516bd8c53463b51e3fb4d947d43f7515d84ec73&vid=9516bd8c53463b51e3fb4d947d43f7515d84ec73&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR