(Trifluoromethyl)trimethylsilane ≥95.0%

Supplier: TCI America

Synonyms: trimethyl(trifluoromethyl)silane

T1570-25G T1570-5G
TCT1570-25GEA 111.56 USD
TCT1570-25G TCT1570-5G
(Trifluoromethyl)trimethylsilane ≥95.0%
(Trifluoromethyl)trimethylsilane

[Trifluoromethylating Reagent]
CAS Number: 81290-20-2
MDL Number: MFCD00145454
Molecular Formula: C4H9F3Si
Molecular Weight: 142.20
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 55
Flash Point (°C): -32
Specific Gravity (20/20): 0.96


Product Categories: Synthetic Organic Chemistry, Fluorinating Agents, Building Blocks, Fluorinated Bioactive Compounds, Difluoromethylating Agents, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: (Trimethylsilyl)trifluoromethane, Ruppert-Prakash Reagent

Formula: C₄H₉F₃Si
Boiling Pt: 55 °C
Density: 0.96 (20 °C)
Flash Pt: –32 °C
MDL Number: MFCD00145454
CAS Number: 81290-20-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5094a4484685a40f5a4d2dded39ab607fc02661&vid=6bde07bf8d968da54f30e69af8a439116c68f7cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR