3,3,5,5-Tetramethyl-1-pyrroline-N-oxide ≥98.0% (by GC)

Supplier: TCI America
Warning

T1554-1G T1554-100MG
TCT1554-1GEA 436.2 USD
TCT1554-1G TCT1554-100MG
3,3,5,5-Tetramethyl-1-pyrroline-N-oxide ≥98.0% (by GC)
3,3,5,5-Tetramethyl-1-pyrroline-N-oxide

CAS Number: 10135-38-3
MDL Number: MFCD00010542
Molecular Formula: C8H15NO
Molecular Weight: 141.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 73
Melting point (°C): 61
Storage Temperature: 0-10°C

Formula: C₈H₁₅NO
MW: 141.21 g/mol
Boiling Pt: 73 °C
Melting Pt: 61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010542
CAS Number: 10135-38-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8db82834d6c5b7980f88b0264d3a86c8a84503b6&vid=4ba5fa471a19d772a4cd464a75560094fa1c7cd2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 62.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR