1,4-Thioxane ≥97.0%

Supplier: TCI America

Synonyms: 1,4-Oxathiane

T1543-25G T1543-5G
TCT1543-25GEA 95 USD
TCT1543-25G TCT1543-5G
1,4-Thioxane ≥97.0%
1,4-Thioxane

CAS Number: 15980-15-1
MDL Number: MFCD00006573
Molecular Formula: C4H8OS
Molecular Weight: 104.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 67
Flash Point (°C): 42
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 1,4-Oxathiane

Formula: C₄H₈OS
MW: 104.17 g/mol
Boiling Pt: 67 °C
Density: 1.12 (20 °C)
Flash Pt: 42 °C
MDL Number: MFCD00006573
CAS Number: 15980-15-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75c7daa793a83b641582aa7222d434a11d225838&vid=75c7daa793a83b641582aa7222d434a11d225838&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR