Triisopropylsilane ≥98.0%

Supplier: TCI America

T1533-100ML T1533-25ML T1533-5ML
TCT1533-100MLEA 166.52 USD
TCT1533-100ML TCT1533-25ML TCT1533-5ML
Triisopropylsilane ≥98.0%
Triisopropylsilane

CAS Number: 6485-79-6
MDL Number: MFCD00009657
Molecular Formula: C9H22Si
Molecular Weight: 158.36
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 166
Flash Point (°C): 37
Specific Gravity (20/20): 0.77

Formula: C₉H₂₂Si
Boiling Pt: 166 °C
Density: 0.77 (20 °C)
Flash Pt: 37 °C
MDL Number: MFCD00009657
CAS Number: 6485-79-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=476a2193bc8767e9dbe4c499f615b0630f878d01&vid=476a2193bc8767e9dbe4c499f615b0630f878d01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR