tert-Butyldimethylsilyl trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
t-Butyldimethylsilyl trifluoromethanesulphonate
CAS Number: 69739-34-0
MDL Number: MFCD00000405
Molecular Formula: C7H15F3O3SSi
Molecular Weight: 264.33
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 36
Specific Gravity (20/20): 1.15
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Protection & Derivatization Reagents (for Synthesis), Silicon Compounds (for Synthesis), Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: tert-Butyldimethylsilyl Triflate, TBDMS Triflate, Trifluoromethanesulfonic Acid tert-Butyldimethylsilyl Ester, TBS Triflate
Formula:
C₇H₁₅F₃O₃SSi MW: 264.34 g/mol Density: 1.15 (20 °C) Flash Pt: 36 °C |
MDL Number:
MFCD00000405 CAS Number: 69739-34-0 UN: 2920 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...