2-(Trifluoromethyl)acrylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 381-98-6
MDL Number: MFCD00042424
Molecular Formula: C4H3F3O2
Molecular Weight: 140.06
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 148
Melting point (°C): 52
Flash Point (°C): 82
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Monomers
Other Product Names / Synonyms: 2-(Trifluoromethyl)propenoic Acid
Formula:
C₄H₃F₃O₂ Boiling Pt: 148 °C Melting Pt: 52 °C Flash Pt: 82 °C |
MDL Number:
MFCD00042424 CAS Number: 381-98-6 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...