Toluene-p-sulfonic anhydride ≥95.0% (by titrimetric analysis)
Supplier: TCI America
T1485-25G
T1485-5G
TCT1485-25GEA
135
USD
TCT1485-25G
TCT1485-5G
Toluene-p-sulfonic anhydride ≥95.0% (by titrimetric analysis)
Toluene-p-sulphonic anhydride
CAS Number: 4124-41-8
MDL Number: MFCD00008548
Molecular Formula: C14H14O5S2
Molecular Weight: 326.38
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 125
Formula:
C₁₄H₁₄O₅S₂ Melting Pt: 125 °C |
MDL Number:
MFCD00008548 CAS Number: 4124-41-8 UN: 3261 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...