m-Tolualdehyde-2,4-dinitrophenylhydrazone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3'-Methylbenzaldehyde 2,4-Dinitrophenylhydrazone

T1448-10G T1448-1G
TCT1448-10GEA 258 USD
TCT1448-10G TCT1448-1G
m-Tolualdehyde-2,4-dinitrophenylhydrazone ≥98.0% (by titrimetric analysis)
m-Tolualdehyde-2,4-dinitrophenylhydrazone

CAS Number: 2880-05-9
MDL Number: MFCD00191585
Molecular Formula: C14H12N4O4
Molecular Weight: 300.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Yellow
Melting point (°C): 213


Other Product Names / Synonyms: 3'-Methylbenzaldehyde 2,4-Dinitrophenylhydrazone

Formula: C₁₄H₁₂N₄O₄
MW: 300.27 g/mol
Melting Pt: 213 °C
MDL Number: MFCD00191585
CAS Number: 2880-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d0b42f777a1ee4390df44a030b40cbde131dec0&vid=4d0b42f777a1ee4390df44a030b40cbde131dec0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR