o-Tolualdehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2'-Methylbenzaldehyde 2,4-Dinitrophenylhydrazone

T1447-10G T1447-1G
TCT1447-10GEA 489 USD
TCT1447-10G TCT1447-1G
o-Tolualdehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
o-Tolualdehyde 2,4-Dinitrophenylhydrazone

CAS Number: 1773-44-0
MDL Number: MFCD00191584
Molecular Formula: C14H12N4O4
Molecular Weight: 300.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Yellow
Melting point (°C): 196


Other Product Names / Synonyms: 2'-Methylbenzaldehyde 2,4-Dinitrophenylhydrazone

Formula: C₁₄H₁₂N₄O₄
MW: 300.27 g/mol
Melting Pt: 196 °C
MDL Number: MFCD00191584
CAS Number: 1773-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78edb6f67591aef658957c409edf53de98b24b2f&vid=78edb6f67591aef658957c409edf53de98b24b2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR