1,1,3,3-Tetramethyldisiloxane ≥97.0% (by GC)

Supplier: TCI America

T1437-250ML T1437-25ML
TCT1437-250MLEA 157.98 USD
TCT1437-250ML TCT1437-25ML
1,1,3,3-Tetramethyldisiloxane ≥97.0% (by GC)
1,1,3,3-Tetramethyldisiloxane

CAS Number: 3277-26-7
MDL Number: MFCD00008256
Molecular Formula: C4H14OSi2
Molecular Weight: 134.33
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 71
Flash Point (°C): -20
Specific Gravity (20/20): 0.76


Product Categories: Synthetic Organic Chemistry, Reduction
Other Product Names / Synonyms: Bis(dimethylsilyl) Ether

Formula: C₄H₁₄OSi₂
MW: 134.33 g/mol
Boiling Pt: 71 °C
Density: 0.76 (20 °C)
Flash Pt: –20 °C
MDL Number: MFCD00008256
CAS Number: 3277-26-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cc5a7b6f2b000a2a83a778934719c037801925e&vid=2cc5a7b6f2b000a2a83a778934719c037801925e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR