2,4,6-Trichlorobenzoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,4,6-Trichlorobenzoyl Chloride

T1413-25G T1413-5G
TCT1413-25GEA 380 USD
TCT1413-25G TCT1413-5G
2,4,6-Trichlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
2,4,6-Trichlorobenzoyl chloride

CAS Number: 4136-95-2
MDL Number: MFCD00075323
Molecular Formula: C7H2Cl4O
Molecular Weight: 243.89
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 275
Flash Point (°C): 113
Specific Gravity (20/20): 1.56

Formula: C₇H₂Cl₄O
MW: 243.9 g/mol
Boiling Pt: 275 °C
Density: 1.56 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00075323
CAS Number: 4136-95-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbb1eff4ca73133aabef39ab0552b7c38863529f&vid=fbb1eff4ca73133aabef39ab0552b7c38863529f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR