4-(Trifluoromethoxy)aniline ≥98.0%

Supplier: TCI America

T1345-25G T1345-5G
TCT1345-25GEA 56 USD
TCT1345-25G TCT1345-5G
4-(Trifluoromethoxy)aniline ≥98.0%
4-(Trifluoromethoxy)aniline

CAS Number: 461-82-5
MDL Number: MFCD00041314
Molecular Formula: C7H6F3NO
Molecular Weight: 177.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 83
Flash Point (°C): 81
Specific Gravity (20/20): 1.32


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha-Trifluoro-p-anisidine

Formula: C₇H₆F₃NO
Boiling Pt: 83 °C
Density: 1.32 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00041314
CAS Number: 461-82-5
UN: 2810
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72cc4c86ac8f99162d8ce4ca0161c546d249be73&vid=72cc4c86ac8f99162d8ce4ca0161c546d249be73&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR