Tris(dimethylamino)phosphine ≥96.0% (by GC) (may contain precipitate)

Supplier: TCI America

Synonyms: Tris(dimethylamino)phosphine, Hexamethyl phosphorous triamide, Hexamethyltriaminophosphine, HMPT

T1317-250ML T1317-25ML
TCT1317-250MLEA 1722 USD
TCT1317-250ML TCT1317-25ML
Tris(dimethylamino)phosphine ≥96.0% (by GC) (may contain precipitate)
Tris(dimethylamino)phosphine

CAS Number: 1608-26-0
MDL Number: MFCD00008301
Molecular Formula: C6H18N3P
Molecular Weight: 163.20
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 163
Flash Point (°C): 26
Specific Gravity (20/20): 0.90
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Hexamethylphosphorous Triamide, HMPT

Formula: C₆H₁₈N₃P
MW: 163.2 g/mol
Boiling Pt: 163 °C
Density: 0.90 (20 °C)
Flash Pt: 26 °C
MDL Number: MFCD00008301
CAS Number: 1608-26-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=718cd87c215892d3e08752d6f59603e45d587cc4&vid=718cd87c215892d3e08752d6f59603e45d587cc4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR