α'-Bromo-α,α,α-trifluoro-p-xylene ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 402-49-3
MDL Number: MFCD00000403
Molecular Formula: C8H6BrF3
Molecular Weight: 239.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 66
Flash Point (°C): 88
Freezing point (°C): 31
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-(Bromomethyl)benzotrifluoride, alpha-Bromo-alpha',alpha',alpha'-trifluoro-p-xylene
Formula:
C₈H₆BrF₃ Boiling Pt: 66 °C Flash Pt: 88 °C |
MDL Number:
MFCD00000403 CAS Number: 402-49-3 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...