Magnesium trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Magnesium trifluoromethanesulphonate, Magnesium triflate
T1304-25G
T1304-5G
TCT1304-25GEA
158
USD
TCT1304-25G
TCT1304-5G
Magnesium trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)
Magnesium trifluoromethanesulphonate
CAS Number: 60871-83-2
MDL Number: MFCD00042601
Molecular Formula: CHF3O3S
Molecular Weight: 322.43
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts
Other Product Names / Synonyms: Magnesium Triflate, Trifluoromethanesulfonic Acid Magnesium Salt
Formula:
C₂F₆MgO₆S₂ MW: 322.45 g/mol |
MDL Number:
MFCD00042601 CAS Number: 60871-83-2 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...