Lanthanum(III) trifluoromethanesulphonate ≥98.0%
Supplier: TCI America
Synonyms:
Lanthanum(III) trifluoromethanesulphonate, Lanthanum trifluoromethanesulphonate, Lanthanum triflate
CAS Number: 52093-26-2
MDL Number: MFCD00077560
Molecular Formula: CHF3O3S
Molecular Weight: 586.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts, Lewis Acids (Multi-component Reaction), Stable Lewis Acids in Aqueous Media
Other Product Names / Synonyms: Lanthanum(III) Triflate, Trifluoromethanesulfonic Acid Lanthanum(III) Salt
Formula:
C₃F₉LaO₉S₃ MW: 586.12 g/mol |
MDL Number:
MFCD00077560 CAS Number: 52093-26-2 UN: 3261 ADR: 8 III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...