2-Butynoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
T1199-1G
T1199-25G
T1199-5G
TCT1199-1GEA
39
USD
TCT1199-1G
TCT1199-25G
TCT1199-5G
2-Butynoic acid ≥98.0% (by GC, titration analysis)
2-Butynoic acid
CAS Number: 590-93-2
MDL Number: MFCD00004363
Molecular Formula: C4H4O2
Molecular Weight: 84.07
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 203
Melting point (°C): 77
Other Product Names / Synonyms: 2-Butynoic Acid
Formula:
C₄H₄O₂ MW: 84.07 g/mol Boiling Pt: 203 °C Melting Pt: 77 °C |
MDL Number:
MFCD00004363 CAS Number: 590-93-2 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...