Ethyl trifluoromethanesulphonate ≥98.0%

Supplier: TCI America

T1191-25G T1191-5G
TCT1191-25GEA 199 USD
TCT1191-25G TCT1191-5G
Ethyl trifluoromethanesulphonate ≥98.0%
Ethyl trifluoromethanesulphonate

CAS Number: 425-75-2
MDL Number: MFCD00000410
Molecular Formula: C3H5F3O3S
Molecular Weight: 178.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 115
Flash Point (°C): 35
Specific Gravity (20/20): 1.38


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: Ethyl Triflate, Trifluoromethanesulfonic Acid Ethyl Ester

Formula: C₃H₅F₃O₃S
MW: 178.13 g/mol
Boiling Pt: 115
Density: 1.38 (20 °C)
Flash Pt: 35 °C
MDL Number: MFCD00000410
CAS Number: 425-75-2
UN: 2920
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28052bab0b320a5c2632a7a04fe54e55a1a763eb&vid=28052bab0b320a5c2632a7a04fe54e55a1a763eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR