Tritylium tetrafluoroborate ≥98.0%
Supplier: TCI America
Synonyms:
Triphenylcarbenium tetrafluoroborate
T1173-25G
T1173-5G
TCT1173-25GEA
423.7
USD
TCT1173-25G
TCT1173-5G
Tritylium tetrafluoroborate ≥98.0%
Tritylium tetrafluoroborate
CAS Number: 341-02-6
MDL Number: MFCD00013120
Molecular Formula: C19H15BF4
Molecular Weight: 330.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Yellow
Melting point (°C): 202
Product Categories: Synthetic Organic Chemistry, Glycosylation
Other Product Names / Synonyms: Triphenylcarbenium Tetrafluoroborate, Tritylium Tetrafluoroborate
Formula:
C₁₉H₁₅BF₄ Melting Pt: 202 °C |
MDL Number:
MFCD00013120 CAS Number: 341-02-6 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...