1-Amino-2,2,2-trifluoroethane ≥97.0% (by GC, titration analysis)

Supplier: TCI America

T1169-25G T1169-5G
TCT1169-25GEA 491 USD
TCT1169-25G TCT1169-5G
1-Amino-2,2,2-trifluoroethane ≥97.0% (by GC, titration analysis)
1-Amino-2,2,2-trifluoroethane

CAS Number: 753-90-2
MDL Number: MFCD00008132
Molecular Formula: C2H4F3N
Molecular Weight: 99.06
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 37
Flash Point (°C): -17
Specific Gravity (20/20): 1.26
Storage Temperature: 0-10°C

Formula: C₂H₄F₃N
Boiling Pt: 37 °C
Density: 1.26 (20 °C)
Flash Pt: –17 °C
MDL Number: MFCD00008132
CAS Number: 753-90-2
UN: 2733
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0b9308d9836453c02f0cd7b31d9b540ea1fd4ce&vid=b0b9308d9836453c02f0cd7b31d9b540ea1fd4ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR