3-Thienylacetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Thiophene-3-acetic acid, 3-Thiopheneacetic acid

T1152-25G T1152-5G
TCT1152-25GEA 351 USD
TCT1152-25G TCT1152-5G
3-Thienylacetic acid ≥98.0% (by titrimetric analysis)
3-Thienylacetic acid

CAS Number: 6964-21-2
MDL Number: MFCD00005473
Molecular Formula: C6H6O2S
Molecular Weight: 142.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 129
Melting point (°C): 80


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 3-Thienylacetic Acid

Formula: C₆H₆O₂S
MW: 142.18 g/mol
Boiling Pt: 129 °C
Melting Pt: 80 °C
MDL Number: MFCD00005473
CAS Number: 6964-21-2
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8281f30f0bd4762485be4aeaa687c844dfa79772&vid=8281f30f0bd4762485be4aeaa687c844dfa79772&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR