1-(Trimethylsilyl)-1-propyne ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 6224-91-5
MDL Number: MFCD00009271
Molecular Formula: C6H12Si
Molecular Weight: 112.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 100
Flash Point (°C): -4
Specific Gravity (20/20): 0.75
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis), Materials Chemistry, Langmuir-Blodgett(LB) Film Forming Agents
Other Product Names / Synonyms: 1-TMS-1-propyne
Formula:
C₆H₁₂Si Boiling Pt: 100 °C Density: 0.75 (20 °C) Flash Pt: –4 °C |
MDL Number:
MFCD00009271 CAS Number: 6224-91-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...