Tropylium tetrafluoroborate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Tropyliumtetrafluoroborate, Cyclohepta-2,4,6-trienylium tetrafluoroborate
T1121-1G
T1121-5G
TCT1121-1GEA
67.09
USD
TCT1121-1G
TCT1121-5G
Tropylium tetrafluoroborate ≥98.0% (by titrimetric analysis)
Tropylium tetrafluoroborate
CAS Number: 27081-10-3
MDL Number: MFCD00013402
Molecular Formula: C7H7BF4
Molecular Weight: 177.94
Purity/Analysis Method:
Formula:
C₇H₇BF₄ MW: 177.94 g/mol |
MDL Number:
MFCD00013402 CAS Number: 27081-10-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...