Trimethylsilyl isocyanate ≥95.0% (by GC)

Supplier: TCI America

T1106-25G T1106-5G
TCT1106-25GEA 201 USD
TCT1106-25G TCT1106-5G
Trimethylsilyl isocyanate ≥95.0% (by GC)
Trimethylsilyl isocyanate

CAS Number: 1118-02-1
MDL Number: MFCD00001993
Molecular Formula: C4H9NOSi
Molecular Weight: 115.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 90
Flash Point (°C): -2
Specific Gravity (20/20): 0.87
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Isocyanatotrimethylsilane

Formula: C₄H₉NOSi
Boiling Pt: 90 °C
Density: 0.87 (20 °C)
Flash Pt: -2 °C
MDL Number: MFCD00001993
CAS Number: 1118-02-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8209b8b5185edc5cb8db9f16af2069f08f1c1d47&vid=8209b8b5185edc5cb8db9f16af2069f08f1c1d47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR