Trifluoromethanesulfonic anhydride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Triflic anhydride
T1100-10G
T1100-250G
T1100-25G
TCT1100-10GEA
90.17
USD
TCT1100-10G
TCT1100-250G
TCT1100-25G
Trifluoromethanesulfonic anhydride ≥98.0% (by titrimetric analysis)
Trifluoromethanesulphonic anhydride
CAS Number: 358-23-6
MDL Number: MFCD00000408
Molecular Formula: C2F6O5S2
Molecular Weight: 282.13
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 84
Melting point (°C): -80
Specific Gravity (20/20): 1.72
Storage Temperature: 0-10°C
Formula:
C₂F₆O₅S₂ MW: 282.14 g/mol Boiling Pt: 84 °C Melting Pt: –80 °C Density: 1.72 (20 °C) |
MDL Number:
MFCD00000408 CAS Number: 358-23-6 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...