Trichloroacetyl isocyanate ≥97.0% (by total nitrogen basis)

Supplier: TCI America

T1047-1G T1047-25G T1047-5G
TCT1047-1GEA 54 USD
TCT1047-1G TCT1047-25G TCT1047-5G
Trichloroacetyl isocyanate ≥97.0% (by total nitrogen basis)
Trichloroacetyl isocyanate

CAS Number: 3019-71-4
MDL Number: MFCD00002033
Molecular Formula: C3Cl3NO2
Molecular Weight: 188.39
Purity/Analysis Method: >97.0% (N)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 85
Flash Point (°C): 65
Specific Gravity (20/20): 1.59
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Isocyanic Acid Trichloroacetyl Ester

Formula: C₃Cl₃NO₂
Boiling Pt: 85 °C
Density: 1.59 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00002033
CAS Number: 3019-71-4
UN: 2927
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1bfa9a63b4d80b011e1c3452acb1e9d642d93bb&vid=e1bfa9a63b4d80b011e1c3452acb1e9d642d93bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR