(p-Toluenesulfonyl)methyl isocyanide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 1-((Isocyanomethyl)sulphonyl)-4-methylbenzene, Tosylmethyl isocyanide

T1046-25G T1046-5G
TCT1046-25GEA 111 USD
TCT1046-25G TCT1046-5G
(p-Toluenesulfonyl)methyl isocyanide ≥98.0% (by HPLC, total nitrogen)
(p-Toluenesulphonyl)methyl isocyanide

CAS Number: 36635-61-7
MDL Number: MFCD00000005
Molecular Formula: C9H9NO2S
Molecular Weight: 195.24
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 110


Product Categories: Synthetic Organic Chemistry, Sulfones, Isonitriles (Multi-component Reaction), Carbon Homologation / Degradation Reaction, Cyanation
Other Product Names / Synonyms: TosMIC, Tosylmethyl Isocyanide

Formula: C₉H₉NO₂S
MW: 195.24 g/mol
Melting Pt: 110 °C
MDL Number: MFCD00000005
CAS Number: 36635-61-7
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0007398119e07782b311d52fad0f6fb77065467a&vid=0007398119e07782b311d52fad0f6fb77065467a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR