1,3,5-Trifluorobenzene ≥98.0% (by GC)

Supplier: TCI America

T1039-25G T1039-5G
TCT1039-25GEA 248 USD
TCT1039-25G TCT1039-5G
1,3,5-Trifluorobenzene ≥98.0% (by GC)
1,3,5-Trifluorobenzene

CAS Number: 372-38-3
MDL Number: MFCD00000333
Molecular Formula: C6H3F3
Molecular Weight: 132.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 76
Flash Point (°C): -7
Specific Gravity (20/20): 1.28

Formula: C₆H₃F₃
MW: 132.09 g/mol
Boiling Pt: 76 °C
Density: 1.28 (20 °C)
Flash Pt: –7 °C
MDL Number: MFCD00000333
CAS Number: 372-38-3
UN: 2387
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=501fbb59acb6be09de344faf1e3bc8960b4d45d1&vid=40ae826d8800e4662a264365ebe88e06226602ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR