Tetrabutylammonium fluoride hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tetrabutylammonium fluoride xhydrate

T1037-100G T1037-25G
TCT1037-100GEA 405.87 USD
TCT1037-100G TCT1037-25G
Tetrabutylammonium fluoride hydrate ≥98.0% (by titrimetric analysis)
Tetrabutylammonium fluoride hydrate

[for Catalyst of acylation, silylation and cleavage of silyl ether]
CAS Number: 22206-57-1
MDL Number: MFCD00011747
Molecular Formula: C16H36FN
Molecular Weight: 261.47
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Fluorinating Agents
Other Product Names / Synonyms: TBAF

Formula: C₁₆H₃₆FN·H₂O
MW: 279.4774 g/mol
MDL Number: MFCD00149980
CAS Number: 22206-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d1a68e48d5d7af9e206fbdbfef15608ec42b6d0&vid=e01b3155302ebba9a8122dff2d77cb15e85d8e6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR