Thiomorpholine ≥98.0% (by GC)
Supplier: TCI America
T1007-25ML
T1007-5ML
TCT1007-25MLEA
162
USD
TCT1007-25ML
TCT1007-5ML
Thiomorpholine ≥98.0% (by GC)
Thiomorpholine
CAS Number: 123-90-0
MDL Number: MFCD00005974
Molecular Formula: C4H9NS
Molecular Weight: 103.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 169
Flash Point (°C): 63
Specific Gravity (20/20): 1.09
Other Product Names / Synonyms: Thiamorpholine
Formula:
C₄H₉NS MW: 103.19 g/mol Boiling Pt: 169 °C Density: 1.09 (20 °C) Flash Pt: 63 °C |
MDL Number:
MFCD00005974 CAS Number: 123-90-0 UN: 3267 ADR: 8,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...