Tetrabutylammonium tetrahydroborate ≥96.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tetra-n-butylammonium borohydride, Tetrabutylammonium borohydride, Tetrabutylammonium tetrahydridoborate

T0917-25G T0917-5G
TCT0917-25GEA 95.6 USD
TCT0917-25G TCT0917-5G
Tetrabutylammonium tetrahydroborate ≥96.0% (by titrimetric analysis)
Tetrabutylammonium tetrahydroborate

[Reducing Reagent]
CAS Number: 33725-74-5
MDL Number: MFCD00012035
Molecular Formula: C16H40BN
Molecular Weight: 257.31
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: White
Melting point (°C): 126

Formula: C₁₆H₄₀BN
MW: 257.31 g/mol
Melting Pt: 126 °C
MDL Number: MFCD00012035
CAS Number: 33725-74-5
UN: 3131
ADR: 4.3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5e42f005e030ef2df50693950dcd5fb352f0c60&vid=48afed30cb3ed0307ec7ebe8868010fca582e2f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR