Trimethylsilyl trifluoromethanesulphonate ≥98.0%
Supplier: TCI America
Synonyms:
Trimethylsilyl trifluoromethanesulphonate
[Trimethylsilylating Agent]
CAS Number: 27607-77-8
MDL Number: MFCD00000406
Molecular Formula: C4H9F3O3SSi
Molecular Weight: 222.25
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 140
Flash Point (°C): 25
Specific Gravity (20/20): 1.23
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Glycosylation, Protection & Derivatization Reagents (for Synthesis), Nucleic Acid Synthesis, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Trifluoromethanesulfonic Acid Trimethylsilyl Ester, Trimethylsilyl Triflate, TMSOTf
Formula:
C₄H₉F₃O₃SSi MW: 222.26 g/mol Boiling Pt: 77 Density: 1.23 (20 °C) Flash Pt: 25 °C |
MDL Number:
MFCD00000406 CAS Number: 27607-77-8 Merck Index: 12,09849 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...