Tetramethylammonium tetrahydroborate ≥90.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tetramethylammonium tetrahydridoborate, Tetramethylammonium borohydride

T0852-25G T0852-5G
TCT0852-25GEA 100 USD
TCT0852-25G TCT0852-5G
Tetramethylammonium tetrahydroborate ≥90.0% (by titrimetric analysis)
Tetramethylammonium tetrahydroborate

[Reducing Reagent]
CAS Number: 16883-45-7
MDL Number: MFCD00011778
Molecular Formula: C4H16BN
Molecular Weight: 88.99
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Color: White

Formula: C₄H₁₆BN
MW: 88.99 g/mol
MDL Number: MFCD00011778
CAS Number: 16883-45-7
UN: 3131
ADR: 4.3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b947ebd6c161e9aa250ef4dd8f4171384474d9a5&vid=9c5c6f73854d00a6a724ef6d62ccc2be8fe26b9f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR