Tetraethylammonium tetrahydroborate ≥90.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tetraethylammonium borohydride, Tetraethylammonium tetrahydridoborate

T0837-25G T0837-5G
TCT0837-25GEA 220.77 USD
TCT0837-25G TCT0837-5G
Tetraethylammonium tetrahydroborate ≥90.0% (by titrimetric analysis)
Tetraethylammonium tetrahydroborate

CAS Number: 17083-85-1
MDL Number: MFCD00031600
Molecular Formula: C8H24BN
Molecular Weight: 145.10
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Color: White

Formula: C₈H₂₄BN
MW: 145.1 g/mol
MDL Number: MFCD00031600
CAS Number: 17083-85-1
UN: 3131
ADR: 4.3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=674019dff4dddd420623dbb68c836cb849fe930b&vid=674019dff4dddd420623dbb68c836cb849fe930b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR