Trifluoromethanesulphonic acid ≥98.0%
Supplier: TCI America
Synonyms:
Trifluoromethanesulphonic acid
T0751-10G
T0751-250G
T0751-25G
TCT0751-10GEA
45.25
USD
TCT0751-10G
TCT0751-250G
TCT0751-25G
Trifluoromethanesulphonic acid ≥98.0%
Trifluoromethanesulphonic acid
CAS Number: 1493-13-6
MDL Number: MFCD00007514
Molecular Formula: CHF3O3S
Molecular Weight: 150.07
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 162
Freezing point (°C): -40
Specific Gravity (20/20): 1.71
Product Categories: Synthetic Organic Chemistry, Glycosylation
Other Product Names / Synonyms: Perfluoromethanesulfonic Acid, Triflic Acid
Formula:
CHF₃O₃S MW: 150.08 g/mol Boiling Pt: 162 Melting Pt: 35 Density: 1.71 (20 °C) |
MDL Number:
MFCD00007514 CAS Number: 1493-13-6 UN: 3265 ADR: 8 II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...