1-(Trifluoroacetyl)imidazole ≥98.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 1546-79-8
MDL Number: MFCD00014501
Molecular Formula: C5H3F3N2O
Molecular Weight: 164.09
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 83
Flash Point (°C): 45
Storage Temperature: 0-10°C
Formula:
C₅H₃F₃N₂O Boiling Pt: 83 °C Flash Pt: 45 °C |
MDL Number:
MFCD00014501 CAS Number: 1546-79-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...