Triethylsilane ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 617-86-7
MDL Number: MFCD00009018
Molecular Formula: C6H16Si
Molecular Weight: 116.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 108
Flash Point (°C): -3
Specific Gravity (20/20): 0.73
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...