2,4,6-Triisopropylbenzenesulfonyl chloride ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2,4,6-Triisopropylbenzene-1-sulphonyl chloride
T0459-25G
T0459-500G
TCT0459-25GEA
22
USD
TCT0459-25G
TCT0459-500G
2,4,6-Triisopropylbenzenesulfonyl chloride ≥97.0% (by titrimetric analysis)
2,4,6-Triisopropylbenzenesulphonyl chloride
CAS Number: 6553-96-4
MDL Number: MFCD00007433
Molecular Formula: C15H23ClO2S
Molecular Weight: 302.86
Purity/Analysis Method: >97.0% (T)
Form: Crystalline Powder
Color: White
Melting point (°C): 96
Storage Temperature: 0-10°C
Formula:
C₁₅H₂₃ClO₂S MW: 302.87 g/mol Melting Pt: 96 °C |
MDL Number:
MFCD00007433 CAS Number: 6553-96-4 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...